Learn More
Enzo Life Sciences Noxa monoclonal antibody (114C307.1) (100 g)
Supplier: Enzo ALX804408C100
Clone: 114C307.1. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human Noxa. UniProt: Q13794. Species Reactivity: Human, Mouse. Applications: ICC, IHC (PS), WB. Recommended dilutions: Immunohistochemistry (paraffin sections, 5g/ml). Western Blot (1-2g/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.05% BSAand0.05% sodium azide. Handling: Avoid freeze/thaw cycles.