Learn More
Solstice NOWPak™ liner-based bottle system, Accessory, Pressure Dispense Connector
Catalog No. 6003062
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
6003062 Each
1 options
Catalog No. 6003062 Supplier Solstice Supplier No. NP515
For use with NOWPak™ Containers
This connector allows for general purpose dispensing of solvent from NOWPak™ II containers using up to 15 psig pressure. It consists of: A cap connector with a 4in. PFA elbow solvent fitting, a 4in. inert gas elbow pressure fitting and a 15 psig pressure relief valve. An FEP diptube with a 40-micron stainless steel frit to prevent drips while changing containers. A 10' harness of 4in. FEP solvent tubing and 4in. nylon pressure tubing.
Specifications
Quantity Each