Learn More
Solstice NOWPak™ liner-based bottle system, Accessory, Pressure Dispense Connector, AKTA
Catalog No. 6003061
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
6003061 Each
1 options
Catalog No. 6003061 Supplier Solstice Supplier No. NP538
For use with NOWPak™ Containers
Solvent can be dispensed at pressures less than 15 psig from NOWPak™ containers connected directly to DNA and peptide synthesizers. Each adapter consists of: A pressure dispense cap with PFA solvent fitting, an inert gas pressurization fitting and a 15 psig pressure relief valve. An FEP diptube with a 40-micron stainless steel frit to prevent drips while changing containers.
Specifications
Quantity Each