missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Protein A Native Protein
Description
Native Protein A. The Protein A Native Protein has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
| Product Type | Protein A Protein |
| Quantity | 2 mL |
| Product Line | Novus |
| Concentration | 6 mg/mL |
| For Use With (Application) | Western Blot, Immunoprecipitation |
| Gene ID (Entrez) | 3919448 |
| Gene Symbol | SAOUHSC_00069 |
| Protein Subtype | Protein A |
| Content And Storage | Store at 4°. Do not freeze. |