missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Emerin Antibody Blocking Peptide

Catalog No. NBP176757E
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176757E 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176757E Supplier Novus Biologicals™ Supplier No. NBP176757PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide containing 19 amino acid peptide from near the amino terminus of human Emerin. Source: Synthetic (Accession #: NP_000108) The Emerin Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 2010
Species Human
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-76757. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias emerin, Emery-Dreifuss muscular dystrophy, LEM domain containing 5, STAEDMDLEMD5
Gene Symbol EMD
Product Type Emerin
Quantity 0.05 mg
Regulatory Status RUO
Source Synthetic
Termination 19 amino acid peptide from near the amino terminus of human Emerin
Show More Show Less