missing translation for 'onlineSavingsMsg'
Learn More

Novobiocin Sodium Salt, MP Biomedicals™

Catalog No. ICN15595725
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15595725 25 g
ICN15595705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15595725 Supplier MP Biomedicals Inc Supplier No. 0215595725

Inhibits DNA synthesis by inhibiting the enzyme Topoisomerase II. Novobiocin Sodium Salt, MP Biomedicals is an antibiotic produced by Streptomyces nieveus. It is active against gram-positive bacteria. It is also a Hsp90 inhibitor 3.

Novobiocin inhibits DNA synthesis by inhibiting bacterial DNA gyrase targeting the GyrB subunit of the enzyme involved in energy transduction. It acts as a competitive inhibitor of the ATPase reaction catalysed by GyrB 2. Uses include:

  • For the production of positively supercoiled plasmid DNA
  • Inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase
  • Inhibitor of retrovirus RNA-dependent DNA-polymerase
  • To study heat shock protein inhibition

Chemical Identifiers

CAS 1476-53-5
Molecular Formula C31H35N2NaO11
Molecular Weight (g/mol) 634.61
MDL Number MFCD00066541,MFCD00066541
InChI Key AXOUUAINTJNFRS-UHFFFAOYNA-N
Synonym Albamycin, Cathomycin sodium
PubChem CID 131673945
SMILES [Na+].COC1C(OC(N)=O)C(O)C(OC2=CC=C3C(=O)[C-](NC(=O)C4=CC=C(O)C(CC=C(C)C)=C4)C(=O)OC3=C2C)OC1(C)C

Specifications

Physical Form Powder
Absorbance λ max: 307nm
Melting Point 220°C
Extinction Coefficient E1%= 600 (0.1M NAOH)
Potency ∼900μg/mg
Quantity 25 g
Optical Rotation −38.0° (2.5g/100ml ethanol at 24°C for the free acid)
Solubility Information Freely soluble in water (10%, clear, colorless solution)
IUPAC Name [(3R,4S,5R,6R)-5-hydroxy-6-[4-hydroxy-3-[[4-hydroxy-3-(3-methylbut-2-enyl)benzoyl]amino]-8-methyl-2-oxochromen-7-yl]oxy-3-methoxy-2,2-dimethyloxan-4-yl] carbamate;molecular hydrogen;sodium
Formula Weight 634.6
Recommended Storage Store at 2–8°C.