Learn More
Novobiocin sodium salt, 20 mg/ml in distilled water, sterile-filtered
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Novobiocin sodium salt, first isolated from Streptomyces nieveus, is a topoisomerase II inhibitor. It is a coumarin compound showing antibiotic activity
• Novobiocin sodium salt can inhibit DNA synthesis by inhibiting the topoisomerase II enzyme. Its antibacterial activity arises from competitive inhibition of the ATPase reaction catalyzed by the GyrB subunit
Application
• Novobiocin sodium salt is used to inhibit the activity of retroviral RNA-dependent DNA polymerase in laboratory experiments
• It is also used as an inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase.
• Novobiocin sodium salt shows anti-staphylococcal activity against methicillin-resistant Staphylococcus aureus, a gram-positive bacterium
• It permits the relaxation of negative DNA supercoils and has been used to generate positively supercoiled DNA
• This compound inhibits the chaperone activity of Hsp90
Specifications
Specifications
| Type | Novobiocin Sodium Salt |
| Physical Form | Liquid |
| Quantity | 50 mL |
| IUPAC Name | sodium 7-{[4-(carbamoyloxy)-3-hydroxy-5-methoxy-6,6-dimethyloxan-2-yl]oxy}-3-[4-hydroxy-3-(3-methylbut-2-en-1-yl)benzamido]-8-methyl-2,4-dioxo-3,4-dihydro-2H-1-benzopyran-3-ide |
| Chemical Name or Material | Novobiocin sodium salt, 20mg/mL in distilled water, sterile-filtered |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H317 May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement P261-P280-P363-P321-P333+P313-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. Wash contaminated clothing before reuse. Specific treatment (see label). If skin irritation or rash occurs: Get medical advice/attention. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 216-023-6 |
| RTECSNumber | RD5425000 |
| TSCA | No |
| storageNote1 | -30° to -10°C |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only