missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Zymogram Developing Buffer (10X)

Catalog No. LC2671
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC2671 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC2671 Supplier Invitrogen™ Supplier No. LC2671

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Novex Zymogram Gels

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature
Buffer Developing Buffers
Concentration 10 X
Gel Type Zymogram
Gel Compatibility Novex™ Zymogram Gels
Quantity 500 mL
Shipping Condition Room Temperature
Product Line Novex
Product Type Zymogram Developing Buffer
Does the Colloidal Blue Staining kit work with Invitrogen Zymogram gels?

Yes, the Colloidal Blue Staining kit works very well with Invitrogen Zymogram gels.

What is the Brij 35 detergent concentration in the 1X Zymogram Developing buffer?

The Brij 35 detergent concentration for Zymogram Developing buffer should be 0.02% (w/v).

For Research Use Only. Not for use in diagnostic procedures.