missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZOOM™ IPG Strip, pH 4-7

Catalog No. ZM0012
Encompass
Change view
Click to view available options
Quantity:
12 strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ZM0012 12 strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ZM0012 Supplier Invitrogen™ Supplier No. ZM0012
Only null left

Includes

Each pack of ZOOM™ Strips contains 12 strips attached to a tri-fold card.

Requires

ZOOM IPGRunner System

Take advantage of oil-free, trouble-free isoelectric focusing (IEF)

  • Optimal IEF resolution zooming in on target proteins
  • Fast isoelectric focusing of proteins and easy handling
  • Each ZOOM Strip is 7cm long and contains a thin layer of polyacrylamide gel that includes a linear pH gradient
  • One-unit narrow pH range for expanding protein separation within a specific region of interest
  • Individually labeled with unique identifying numbers, pH range, and orientation marks

2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

Specifications

Content And Storage Each pack of ZOOM™ Strips contains 12 strips attached to a tri-fold card. Store at -20°C. Guaranteed stable for one year when properly stored.
Gel Size Mini
pH Range 4.0 - 7.0
Quantity 12 strips
Shipping Condition Dry Ice
Product Line ZOOM
Product Type IPG Strip

For Research Use Only. Not for use in diagnostic procedures.