missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ StainEase™ Staining Trays

Catalog No. NI2400
Encompass
Change view
Click to view available options
Quantity:
2 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NI2400 2 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NI2400 Supplier Invitrogen™ Supplier No. NI2400

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

StainEase™ Gel Staining Tray comes with two clear polycarbonate containers, two white polycarbonate strainers, two clear polycarbonate lids with handles and waterproof lid labels.

Stains and solutions can be changed without having to handle the gel

The StainEase Gel Staining Tray is a convenient way to evenly stain mini-gels and prevent gels from breaking. The staining tray assembly includes a perforated strainer insert that holds the gel and drains away the stain, a lid, and an outer container. Easily fits up to four mini-gels.

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Contents: StainEase™ Gel Staining Tray comes with two clear polycarbonate containers, two white polycarbonate strainers, two clear polycarbonate lids with handles and waterproof lid labels.
Quantity 2 pack
Shipping Condition Room Temperature
Product Line StainEase
Product Type Staining Tray
Do you offer staining trays for staining protein gels?

We offer the following staining trays for staining protein gels:
- StainEase Staining Trays (Cat. No. NI2400) for convenient and even staining of Mini and Midi gels. The staining tray assembly includes a perforated strainer insert that holds the gel and drains away the stain, a lid, and an outer container in which stains and solutions can be changed without having to handle the gel. Each tray easily fits up to 4 mini gels.
- Mini Gel Incubation Trays (Cat. No. 22843) for staining mini gels and western blot membranes.

For Research Use Only. Not for use in diagnostic procedures.