missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Protein Molecular Weight Standards (broad range)

Catalog No. p6649
Encompass
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P6649 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P6649 Supplier Invitrogen™ Supplier No. P6649
Only null left

Include 11 proteins from 6500 to 205,000 daltons

The broad range protein molecular standards for use as molecular weight standards in SDS-polyacrylamide gel electrophoresis.

  • Novex Tricine Gels, Novex Tris-Glycine Gels, NuPAGE Bis-Tris Gels, NuPAGE Tris-Acetate Gels, SDS-PAGE Gels

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Number of Markers 11
Ready to Load No
Sample Loading Volume 2 μL
Size Range 6.5 to 205 kDa
Gel Compatibility Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol) 205, 116, 97, 80, 66, 55, 45, 30, 21, 14, 6.5 kDa
Quantity 400 μL
Shipping Condition Wet Ice
Product Type Protein Molecular Weight Standard
Stain Type Unstained
System Type SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.