missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Transfer Buffer (20X)

Catalog No. p-4918341
Encompass
Change view
Click to view available options
Quantity:
125mL
1L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP0006 125mL
NP00061 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NP0006 Supplier Invitrogen™ Supplier No. NP0006
Only null left

Use to transfer proteins from NuPAGE Novex gels to membranes for Western blotting

NuPAGE Transfer Buffer can be used with the Novex Semi-Dry Blotter for semi-dry transfer or with the XCell II™ Blot Module for wet (tank) transfer. NuPAGE Antioxidant may be used with NuPAGE Transfer Buffer to enhance transfer of reduced proteins to membranes.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature. Guaranteed stable for 6 months unless otherwise specified in the product literature.
Buffer Transfer Buffers
Concentration 20X
Gel Type NuPAGE
Gel Compatibility NuPAGE™ Gels
Quantity 125mL
Shipping Condition Room Temperature
Product Line NuPAGE
Product Type Transfer Buffer

For Research Use Only. Not for use in diagnostic procedures.