missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Large Protein Blotting Kit DFS Item

Specifically designed for large protein analysis on NuPAGE Novex Tris-Acetate Gels under denaturing conditions

Supplier:  Invitrogen™ LP0001

Catalog No. LP0001


Only null left
Explore available promotions

Description

Description

The HiMark™ Pre-Stained and Unstained High Molecular Weight Protein Standards offer a time-saving load-and-go format. The HiMark Pre-stained Standard includes one pink colored orientation band and eight blue bands ranging in apparent molecular weight of 31 - 460kDa in NuPAGE Tris-Acetate Gel/SDS buffer system. It's suitable for monitoring electrophoresis, western transfer, and approximating molecular weight of unknown big proteins. The HiMark Unstained Standard, along with the NuPAGE Tris-Acetate Gel/SDS buffer system, offers the highest accuracy for molecular weight estimation of large proteins. It consists of nine proteins, ranging in size from 40 to 500 kDa in NuPAGE Novex 3-8% and 7% Tris-Acetate Gel/SDS buffer systems. Visualize with Coomassie™ stain, silver, and fluorescent protein stains post-electrophoresis. Several NuPAGE Large Protein Starter Kits are available to provide you with all necessary reagents and optimized protocols for high-resolution large protein separation and analysis. Each kit includes two boxes of NuPAGE 3-8% Tris-Acetate Gels, Tris-Acetate SDS Buffer Kit, a staining reagent or blotting membranes, and a vial of the appropriate HiMark Standard.

  • Gel Percentage: 3-8 %
  • Gel Size: Mini (8 x 8 cm)
  • Well Format: 1D Well

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting

Specifications

Specifications

See product manual for kit contents and component storage conditions. Components of each kit are also available individually.
3 to 8%
Tris-Acetate
25 μL
Unstained
NuPAGE
1.0 mm
XCell SureLock Mini-Cell
Mini
Protein Blotting Kit
40 to 500 kDa
10-well
20 gels kit
1D Well
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.