missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NativePAGE™ Sample Prep Kit

Catalog No. BN2008
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BN2008 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BN2008 Supplier Invitrogen™ Supplier No. BN2008

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

1mL of 10% DDM, 1mL of 5% Digitonin, 10mL of NativePAGE, 4X Sample Buffer, 0.5mL NativePAGE 5% G-250 Sample Additive

Includes sample preparation reagents for native gel electrophoresis

The kit includes two ready-to-use detergent solutions, 10% DDM (n-dodecyl-β-D-maltoside) and 5% Digitonin, which improve the solubility of hydrophobic and membrane proteins during sample preparation. The samples prepared with 10% DDM, 5% Digitonin, or the NativePAGE Sample Prep Kit are compatible with NativePAGE Novex Bis-Tris Gels for native electrophoresis, showing increased resolution and reduced streaking.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Contains:
• 1 ml of 10% DDM
• 1 ml of 5% Digitonin
• 10 ml of NativePAGE™ 4X Sample Buffer
• 0.5 ml NativePAGE™ 5% G-250 Sample Additive.

Store the kit at 2°C to 8°C.
Buffer Sample Loading Buffers
Gel Type Native-PAGE
Gel Compatibility NativePAGE™ Gels
Quantity 1 kit
Shipping Condition Wet Ice
Product Line NativePAGE
Product Type PAGE Sample Prep Kit
I would like to run a NativePAGE gel. Which of your protein standards should I use?

We recommend using the NativeMark Unstained Protein Standard, Cat. No. LC0725 for native gel electrophoresis with Tris-Glycine, NuPAGE Tris-Acetate or NativePAGE gels.

Can I use the NativePAGE Sample and Running buffers with NuPAGE Bis-Tris gels?

We do not recommend using NativePAGE Sample and Running buffers with NuPAGE Bis-Tris gels. NuPAGE Invitrogen Bis-Tris gels are optimized for denaturing conditions and have an extremely low operating pH (pH 7.0), which makes it difficult for most proteins to migrate through them in a native state.

Can I use the NativePAGE Sample and Running buffers with Invitrogen Tris-Glycine gels or NuPAGE Tris-Acetate gels for native applications?

NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including NuPAGE Tris-Acetate or Invitrogen Tris-Glycine gels. For those gels, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.

Can I use the NativePAGE Sample and Running buffers with Invitrogen Tris-Glycine gels for native applications?

NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including Invitrogen Tris-Glycine gels. For those gels, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.

Can I use the NativePAGE Sample and Running buffers with NuPAGE Tris-Acetate gels for native applications?

NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including NuPAGE Tris-Acetate gels. When running Tris-Acetate gels under native conditions, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.

For Research Use Only. Not for use in diagnostic procedures.