missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NativePAGE™ Sample Buffer (4X)

Catalog No. bn2003
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BN2003 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BN2003 Supplier Invitrogen™ Supplier No. BN2003
Only null left

A solution used to solubilize native protein samples prior to use with the NativePAGE Gel System

The buffer is used along with 10% DDM or 5% Digitonin detergents to solubilize hydrophobic or membrane proteins. This buffer is also included in the NativePAGE Sample Prep Kit. The samples prepared with 10% DDM, 5% Digitonin, or the NativePAGE Sample Prep Kit are compatible with NativePAGE Novex Bis-Tris Gels for native electrophoresis showing increased resolution and reduced streaking.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Store at 2°C to 8°C
Buffer Sample Loading Buffers
Concentration 4 X
Gel Type Native-PAGE
Gel Compatibility NativePAGE™ Gels
Quantity 10 mL
Shipping Condition Wet Ice
Product Line NativePAGE
Product Type PAGE Sample Buffer

For Research Use Only. Not for use in diagnostic procedures.