missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NativePAGE™ Running Buffer (20X)

Catalog No. bn2001
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BN2001 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BN2001 Supplier Invitrogen™ Supplier No. BN2001
Only null left

Used to make the NativePAGE Cathode and Anode Running Buffers

For use with an XCell SureLock™ Mini Cell when running NativePAGE Gels.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature
Buffer Running Buffers
Concentration 20 X
Gel Type Native-PAGE
Gel Compatibility NativePAGE™ Gels
Quantity 1 L
Shipping Condition Room Temperature
Product Line NativePAGE
Product Type PAGE Running Buffer

For Research Use Only. Not for use in diagnostic procedures.