missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NativePAGE™ Cathode Buffer Additive (20X)

Catalog No. BN2002
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BN2002 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BN2002 Supplier Invitrogen™ Supplier No. BN2002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For running NativePAGE Gels

NativePAGE Cathode Buffer Additive (20X) is added to the NativePAGE Running Buffer and the resulting buffer is used in the cathode chamber of an XCell SureLock™ Mini Cell for running NativePAGE Gels.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature
Buffer Anode & Cathode Buffers
Concentration 20 X
Gel Type IEF
Gel Compatibility NativePAGE™ Gels
Quantity 250 mL
Shipping Condition Room Temperature
Product Line NativePAGE
Product Type PAGE Cathode Buffer Additive
What gels can I use to separate native proteins?

The NativePAGE Invitrogen Bis-Tris Gel System is a pre-cast polyacrylamide mini gel system that provides a sensitive and high-resolution method for analyzing native membrane protein complexes, native soluble proteins, molecular mass estimations, and assessing purity of native proteins. It is based on the blue native polyacrylamide gel electrophoresis technique (BN PAGE) developed by Schagger and von Jagow.

After transferring onto a nitrocellulose membrane, what is the best way to store the membrane overnight for probing the next day?

We recommend rinsing the membrane briefly in water, air drying and store it at room temperature in a ziplock bag. Do not place nitrocellulose in the freezer because it will shatter. Unlike PVDF, nitrocellulose membranes should never be pre-wetted in alcohol as it will cause them to shrivel.

I would like to run a NativePAGE gel. Which of your protein standards should I use?

We recommend using the NativeMark Unstained Protein Standard, Cat. No. LC0725 for native gel electrophoresis with Tris-Glycine, NuPAGE Tris-Acetate or NativePAGE gels.

Can I use the NativePAGE Sample and Running buffers with NuPAGE Bis-Tris gels?

We do not recommend using NativePAGE Sample and Running buffers with NuPAGE Bis-Tris gels. NuPAGE Invitrogen Bis-Tris gels are optimized for denaturing conditions and have an extremely low operating pH (pH 7.0), which makes it difficult for most proteins to migrate through them in a native state.

Can I use the NativePAGE Sample and Running buffers with Invitrogen Tris-Glycine gels or NuPAGE Tris-Acetate gels for native applications?

NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including NuPAGE Tris-Acetate or Invitrogen Tris-Glycine gels. For those gels, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.

Can I use the NativePAGE Sample and Running buffers with Invitrogen Tris-Glycine gels for native applications?

NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including Invitrogen Tris-Glycine gels. For those gels, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.

Can I use the NativePAGE Sample and Running buffers with NuPAGE Tris-Acetate gels for native applications?

NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including NuPAGE Tris-Acetate gels. When running Tris-Acetate gels under native conditions, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.


For Research Use Only. Not for use in diagnostic procedures.