missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ IEF Sample Buffer pH 3-10 (2X)

Catalog No. LC5311
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC5311 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC5311 Supplier Invitrogen™ Supplier No. LC5311

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An optimized pre-mixed IEF sample buffer

Novex IEF Sample Buffer pH 3-10 (2X) is optimized pre-mixed IEF sample buffer for using with Novex pH 3-10 IEF gels. IEF is a sensitive technique which is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensure consistent results.

Isoelectric Focusing, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Store in refrigerator (2–8°C).
Buffer Sample Loading Buffers
Concentration 2 X
Gel Type IEF
pH Range 3 to 10
Gel Compatibility Novex™ IEF Gels
Quantity 25 mL
Shipping Condition Wet Ice
Product Line Novex
Product Type IEF Sample Buffer
When staining IEF gels with the Colloidal Blue Staining Kit, is it necessary to use the fixing solution?

Yes, the fix serves two purposes: it fixes the sample in the IEF gel and it helps to remove gel background.

If you do not use the fixing solution, the background on the gels will be high and detection will be less sensitive.

High background is caused by ampholytes remaining in the gel.

For Research Use Only. Not for use in diagnostic procedures.