Learn More
Invitrogen™ IEF Cathode Buffer pH 3-10 (10X)
Description
IEF Cathode Buffer pH 3-10 (10X) is optimized pre-mixed IEF cathode buffer for using with Novex pH 3-10 IEF gels. IEF is a sensitive technique which is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensure consistent results.
Isoelectric Focusing, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Content And Storage | Store in refrigerator (2–8°C). |
| Buffer | Anode & Cathode Buffers |
| Concentration | 10X |
| Gel Type | IEF |
| pH Range | 3 to 10 |
| Gel Compatibility | Novex™ IEF Gels |
| Quantity | 125 mL |
| Shipping Condition | Wet Ice |
| Product Type | IEF Cathode Buffer |
Frequently Asked Questions (FAQs)
Yes, the fix serves two purposes: it fixes the sample in the IEF gel and it helps to remove gel background.
If you do not use the fixing solution, the background on the gels will be high and detection will be less sensitive.
High background is caused by ampholytes remaining in the gel.
Yes, you can use just arginine (either form of arginine) as long as the arginine free base form is used and not the hydrochloric form. Note that the lysine used to make the mixed buffer should be free base lysine; free acid lysine should not be used as a substitute. The reason for adding basic amino acids to the cathode buffer is to increase the ionic strength of the buffer in order to fortify the basic end.
The ZOOM IEF Fractionator includes the following components:
Chamber Assembly Tube with Anode Reservoir
Spill Trough with Cathode Reservoir Lid
Sample Chambers (7)
Sample Chamber O-ring Seals, red (10)
Sample Chamber Caps with O-rings (7)
Cathode End Sealer
Anode End Sealer
Cathode End Screw Cap
Spacers, black (8)
Spares Box 1
Sample Chamber O-ring Seals (8)
Sample Chamber Caps with O-rings (7)
Spares Box 2
Cathode Chamber Seals (2)
Spacers, black (8)
We recommend storing the ZOOM Disks, ZOOM Carrier Ampholytes, ZOOM Focusing buffers, ZOOM Cathode buffer, IEF Anode buffer, and IEF Cathode buffer at 4 degrees C.
For Research Use Only. Not for use in diagnostic procedures.