missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ IEF Anode Buffer (50X) DFS Item

An optimized pre-mixed IEF anode buffer

Supplier:  Invitrogen™ LC5300

Catalog No. LC5300

Only null left
Explore available promotions

Description

Description

Novex IEF Anode Buffer (50X) is optimized pre-mixed IEF anode buffer for using with Novex IEF gels. IEF is a sensitive technique which is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensure consistent results.

Isoelectric Focusing, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store at room temperature.
50 X
IEF Anode Buffer
Novex™
Room Temperature
Anode & Cathode Buffers
IEF
Novex™ IEF Gels
100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.