missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HiMark™ Unstained Protein Standard DFS Item

Designed for analysis of high molecular weight proteins on Novex™ NuPAGE™ Tris-Acetate Gels

Supplier:  Invitrogen™ LC5688

Catalog No. LC5688


Only null left
Explore available promotions

Description

Description

  • Comprehensive - Contains markers with a wide range of high molecular weights, providing nine protein bands in the range of 30-460kDa
  • Convenient - A ready-to-use formulation eliminates the need to heat, reduce, or add sample buffer prior to use
  • Compatible - Intended for use with NuPAGE Novex 3-8% and 7% Tris-Acetate Gels under denaturing conditions
  • Can also be used with NuPAGE Novex 4-12% Bis-Tris Gels with NuPAGE MOPS Running Buffer and Novex 4% Tris-Glycine Gel, however to obtain best results with HMW proteins, always use NuPAGE Novex Tris-Acetate Gels

HiMark High Molecular Weight Protein Standard also available in a pre-stained format.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Contents: one vial of 250 μL each

Storage buffer: 250 mM Tris-HCl (pH 8.5), 0.5 mM EDTA, 50 mM DTT, 10% glycerol, 2% LDS, 0.2 mM Coomassie G-250, and 0.175 mM Phenol Red

Storage: Upon receipt store at -20°C

Yes
40 to 500 kDa
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
250 μL
HiMark
Unstained
9
Tris-acetate gels
Tris-acetate gels
500, 290, 240, 160, 116, 97, 66, 55, 40 kDa
Dry Ice
Protein Ladder
Western Blotting, SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.