missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-PAGE™ SeeBlue™ Pre-stained Standard

Catalog No. LC5700
Encompass
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC5700 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC5700 Supplier Invitrogen™ Supplier No. LC5700

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

500μL (50 applications of 10μL each) of the E-PAGE™ SeeBlue Pre-stained Standard is provided in loading buffer consisting of Tris-HCl, Formamide, and SDS.250μL (50 applications of 5μL each) of the E-PAGE™magicMark™ Unstained Protein Standard is provided in loading buffer consisting of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 1% SDS, and 0.025% Bromophenol Blue

Designed for use with pre-cast E-PAGE™ Electrophoresis System

  • Consists of three blue-dyed and two colored bands ranging in size from 21-261kDa and is suitable for monitoring gel and transfer runs
  • Consists of five recombinant proteins ranging in size from 20-220kDa, each with an IgG binding-site.
  • Can be detected with Coomassie™ stain on the gels or conjugated-antibody directly on western blots
  • Suitable for molecular weight estimation of unknown proteins
  • Gel Compatibility: E-PAGE

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Contents: 500 μL (50 applications of 10 μL each) of the E-PAGE SeeBlue Pre-stained Standard

Storage buffer: Tris-HCl, formamide, and SDS

Storage: Stable for 4 months at +4°C

Detection Method Colorimetric
Number of Markers 5
Ready to Load Yes
Recommended Applications E-PAGE gels
Size Range 21 to 261 kDa
For Use With (Application) E-PAGE gels
Gel Compatibility E-PAGE™ Gels
Molecular Weight (g/mol) 261, 173, 97, 42, 21 kDa
Quantity 500 μL
Shipping Condition Wet Ice
Product Line E-PAGE, SeeBlue
Product Type Protein Ladder
Stain Type Blue, Orange, Pink
System Type E-PAGE
Show More Show Less
I used one of your protein standards for a western transfer and noticed that some of the lower-molecular weight protein bands passed through the membrane. How can I resolve this issue?

- Decrease voltage, current or length of transfer time
- Make sure that the methanol concentration in the transfer buffer is proper; use a methanol concentration of 10-20% methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane.
- Make sure that the SDS concentration (if added) in the transfer buffer is proper, don't use more than 0.02-0.04% SDS. Using too much SDS can prevent binding of proteins to the membrane.
- Check the pore size of the membrane and the size of the target protein. Proteins smaller than 10 kDa will easily pass through a 0.45 µm pore size membrane. If proteins smaller than 10 kDa are of interest, it would be better to use a 0.2 µm pore size membrane.

I used one of your protein standards for a western transfer and noticed that some of the higher-molecular weight bands transferred very poorly to the membrane. Can you offer some tips?

- Increase voltage, current or length of time for transfer
- SDS in the gel and in the SDS-protein complexes promotes elution of the protein from the gels but inhibits binding of the protein to membranes. This inhibition is higher for nitrocellulose than for PVDF. For proteins that are difficult to elute from the gel such as large molecular weight proteins, a small amount of SDS may be added to the transfer buffer to improve transfer. We recommend pre-equilibrating the gel in 2X Transfer buffer (without methanol) containing 0.02-0.04% SDS for 10 minutes before assembling the sandwich and then transferring using 1X transfer buffer containing 10% methanol and 0.01%SDS.
- Methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane, but has some negative effects on the gel itself, leading to a decrease in transfer efficiency. It may cause a reduction in pore size, precipitation of some proteins, and some basic proteins to become positively charged or neutral. Make sure that the methanol concentration in the transfer buffer is not more than 10-20% and that high-quality, analytical grade methanol is used.

I used one of your pre-stained standards on a Tris-Glycine gel and noticed that the molecular weights of the proteins were different than on a NuPAGE Bis-Tris gel. What is the reason for this?

Pre-stained standards have a dye that is covalently bound to each protein that will result in the standard migrating differently in different buffer systems (i.e., different gels). As a result, using a pre-stained standard for molecular weight estimation will only give the apparent molecular weight of the protein. Pre-stained standards may be used for molecular weight approximation, confirming gel migration and estimating blotting efficiency but for accurate molecular weight estimation, an unstained standard should be used.

I used one of your protein standards and am seeing some extra bands in the lane. Can you offer some suggestions?

- While loading, take care to make sure that there is no cross-contamination from adjacent sample lanes.
- Make sure that the correct amount of standard is loaded per lane. Loading too much protein can result in extra bands and this is a problem especially with silver-stained gels.
- Improper storage of the standard or repeated freeze/thawing can result in protein degradation.

I used one of your protein standards and the bands look non-distinct and smeary. What should I do?

Here are some suggestions:

- Make sure that the correct amount of standard is loaded per lane. Loading too much protein can cause smearing and this is a problem especially with silver stained gels.
- Bands will not be as well resolved in low percentage gels. Try using a higher percentage gel.
- If the bands look smeary and non-distinct after a western transfer/detection, this may be due to the antibody being too concentrated. Follow the manufacturer's recommended dilution or determine the optimal antibody concentration by dot-blotting.

A couple of bands in my protein standard are missing on the gel. Can you help me troubleshoot?

Here are some suggestions:

- Check the gel type/percentage of the gel that was used. Depending on the gel type and/or percentage, all the bands may not be seen. For example, the smallest bands of the protein standard may not resolve on a very low percentage gel whereas the higher molecular weight bands may not resolve on a high percentage gel.
- Check the expiration date on the protein standard. Expired lots may result in faded or missing bands due to protein degradation.
- Check the storage conditions for the protein standard. Improper storage conditions will compromise the stability of the proteins in the standard.
- Make sure that the protein standard was not heated/boiled prior to loading on the gel. Our protein standards are ready to load and we do not recommend heating/boiling them as this may cause degradation of proteins in the standard.

What are the apparent molecular weights of the proteins in the E-PAGE SeeBlue Prestained Standard?

The E-PAGE SeeBlue Prestained Standard is designed for use with E-PAGE pre-cast gels, to visualize protein molecular weight ranges during electrophoresis and evaluate western transfer efficiency. The apparent molecular weights of the proteins in the E-PAGE SeeBlue Prestained Standard are shown below:

- Myosin: 261 kDa in 6% E-PAGE gel
- Phosphorylase B: 173 kDa in 6% E-PAGE gel
- BSA: 97 kDa in 6% E-PAGE gel
- Alcohol dehydrogenase: 42 kDa in 6% E-PAGE gel
- Myoglobin: 21 kDa in 6% E-PAGE gel

What are the storage conditions and shelf life for the E-PAGE SeeBlue Prestained Standard?

We recommend storing the E-PAGE SeeBlue Prestained at 4 degrees C and it is stable for 4 months when properly stored.

Do protein standards run differently on a Zymogram gel compared to a regular Tris-Glycine gel?

Zymogram gels are essentially Tris-Glycine gels containing the substrate. Protein standards run based solely on the percentage of acrylamide and hence should run the same in both kinds of gels. It is quite possible though that if the standard is prestained, the proteins will appear a different color because of the staining (or pre-staining) of the Zymogram gels.

Can I use any of your protein standards for estimation of protein quantity (protein quantitation)?

Our protein standards are not designed for protein quantitation.

What is the recommended gel loading volume for your protein standards?

Please find this information in the respective manuals for the individual protein standards.

Do I need to boil your protein standards before loading on the gel?

Our protein standards are ready to load. We do not recommend heating them as this may cause protein degradation.

Do I have to add reducing agent to your protein standards?

Except for our NativeMark Unstained Protein Standard (designed for native electrophoresis), all of the other unstained and prestained standards we offer (Invitrogen Sharp, SeeBlue, SeeBlue Plus2, BenchMark, HiMark) have been pre-reduced (by a proprietary method). Hence, you do not need to add reducing agent.


For Research Use Only. Not for use in diagnostic procedures.