missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E-PAGE™ Loading Buffer 1
Description
- Compatible with E-PAGE gels
- Size: 4.5mL
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Each E-PAGE™ 96 kit includes 8 gels, E-PAGE™ loading buffer 1, 4.5 ml loading buffer, E-PAGE™ buffer 2, 4.5 ml SDS-free loading buffer, 4.5 ml loading buffer, and a Butterfly Opener. The E-Base™ mother and daughter devices and loading buffers are available separately. Store at room temperature. Guaranteed stable for 6 months after manufacturing when stored properly. |
| Buffer | Sample Loading Buffers |
| Gel Type | E-PAGE |
| Gel Compatibility | E-PAGE™ Gels |
| Quantity | 4.5 mL |
| Shipping Condition | Room Temperature |
| Product Line | E-PAGE |
| Product Type | Loading Buffer |
For Research Use Only. Not for use in diagnostic procedures.