Learn More
Invitrogen™ DryEase™ Mini-Gel Drying System DFS Item
For use with DryEase™ Mini-Gel Drying System
Supplier:  Invitrogen™ NI2387
Catalog No. NI2387
Only null left
Explore available promotions
Description
Description
Gel Drying, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
The DryEase™ Mini-Gel Drying Kit contains 2 DryEase™ Mini-Gel Drying Frames with 8 Gel-Dry™ Clamps, a DryEase™ Mini-Gel Drying Base, DryEase™ Mini Cellophane (200 pre-cut sheets), and Gel-Dry™ Drying Solution (500 ml). Store the solution at room temperature. Guaranteed stable for one year when properly stored.
Mini Gel Drying System
1 system
DryEase Mini-Gel Drying System
DryEase, Novex
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.