missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Colloidal Blue Staining Kit

Catalog No. LC6025
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC6025 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC6025 Supplier Invitrogen™ Supplier No. LC6025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

The Colloidal Blue Staining Kit is supplied with sufficient stainer A and stainer B to stain 25 mini-gels

Five times more sensitivity than Coomassie™ R-250— and can detect as little as 10ng of BSA

The Colloidal Blue Staining Kit allows detection of nanogram levels of protein with minimal effort and requires only water to destain. The Colloidal Blue stain uses colloidal chemistry that reduces free dye in solution and improves the protein-to-dye binding ratio. Samples are intensely stained and visible within 3 hours, background staining is virtually eliminated by destaining overnight with water.

  • Easy-to-use, mass spectrometry compatible reagents
  • Great densitometry results

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage The Colloidal Blue Staining Kit is supplied with sufficient stainer A and stainer B to stain 25 mini-gels. Store the kit at room temperature. The kit is guaranteed for 6 months when properly stored unless otherwise stated in the product documentation.
Detection Location In-Gel Detection
Detection Method Colorimetric
Label or Dye Coomassie
Quantity 1 kit
Shipping Condition Room Temperature
Target Molecule Protein
Product Type Protein Gel Stain Kit
Can I use the Colloidal Blue Staining Kit (Cat. No. LC6025) for membrane staining to detect proteins that are transferred to a membrane?

It is not recommended because the background will be too high. Better alternatives include: 1) Invitrogen Reversible Membrane Protein Stain Kit (Cat. No. IB7710).

2) Coomassie (non-colloidal) staining: stain in 0.1% Coomassie Blue R-250 in 50% methanol for 5 min and destain with several changes of 50% methanol and 10% acetic acid. Rinse with several changes of water, air dry and store for up to 12 months at -20°C. Sensitivity is approximately at the 50-100 ng level.

3) Use SimplyBlue SafeStain (Cat. No. LC6060). The SimplyBlue SafeStain manual has the protocol for staining PVDF membranes, but it is not recommended for nitrocellulose because of high background.

4) Amido Black: same as Coomassie but less sensitive.

5) Ponceau S: same as Coomassie but less sensitive.

6) UV transillumination: place membrane on filter paper after blot is finished and allow to dry at room temperature for about 10 min. Rewet in 20% methanol and view the blot in front of white light while it is still wet; the bands will look more translucent than the membrane. If the bands disappear as the membranes dries, rewet again.

Why is Coomassie G-250 used as the tracking dye in NuPAGE LDS Sample Buffer instead of bromophenol blue?

Coomassie G-250 will give a sharp dye front with both NuPAGE MES and MOPS Running Buffers and is therefore used as the tracking dye in the NuPAGE LDS Sample Buffer.

Bromophenol blue runs more slowly than some peptides with the NuPAGE MES Running Buffer system.

Coomassie G-250 migrates much closer to the moving ion front than bromophenol blue, ensuring that small peptides will not be run too far (e.g., off the gel).

Is the SimplyBlue SafeStain the appropriate stain for quantitation by densitometry?

The Colloidal Blue Staining Kit (Cat. No. LC6025) is best for quantitation by densitometry. You can also use SimplyBlue SafeStain for this application.

The great advantage of SimplyBlue SafeStain is that it is very easy to use and safe.

The sensitivity of my SimplyBlue SafeStain seems to be decreasing over time. Why is this?

Check the cap on the bottle. If the bottles are not tightly sealed, the alcohol can evaporate from the stain causing substantial decrease in stain sensitivity.

How do I destain proteins on a PVDF membrane that were stained with SimplyBlue SafeStain?

After staining with SimplyBlue SafeStain, use deionized water for the less strongly retained protein bands on the PVDF membrane.

Increasing methanol or ethanol concentrations up to 70% should destain any remaining bands. You can leave the membrane in the destain indefinitely.

How can membranes be stained?

Membranes cannot be stained with Colloidal Blue, the background will be too high. Better alternatives include:

1) Coomassie (non-colloidal) staining: Stain in 0.1% Coomassie Blue R-250 in 50% methanol for 5 min and destain with several changes of 50% methanol and 10% acetic acid. Rinse with several changes of water, air dry and store for up to 12 months at -20°C. Sensitivity is approximately at the 50-100 ng level.

2) SimplyBlue SafeStain. There is a protocol included in the SimplyBlue manual for staining PVDF but it is not recommended for nitrocellulose because of the high background.

3) Amido Black: less sensitive than Coomassie Blue.
Recipe for amido black: (1L) 450 mL methanol, 450 mL dH2O, 100 mL glacial acetic acid, 0.1 g amido black.
Procedure: combine ingredients and stir until the amido black is dissolved. If the membrane has dried up, pre-wet by floating on dH2O and soaking for 5 min. Transfer to tray containing amido black for 5-10 min. Wash in several changes of dH2O.

4) Ponceau S: less sensitive than Coomassie Blue.
Recipe for Ponceau S (10X stock): 2 g Ponceau S, 30 g trichloroacetic acid, 30 g sulfosalicylic Acid, dH2O to 100 mL
Combine ingredients and stir until Ponceau S is dissolved. Dilute 1:10 before using.
Procedure: If membrane has dried up, pre-wet membrane by floating on dH2O and soaking for 5 min. Transfer membrane to tray containing Ponceau S and incubate for 5-10 min. Wash membrane in several changes of dH2O.

5) UV transillumination: Place membrane on filter paper after blot is finished and allow to dry at room temperature for about 10 min. Rewet in 20% methanol and view the blot in front of white light while it is still wet; the bands will look more translucent than the membrane. If the bands disappear because they dry, rewet again.

What is the sensitivity of Coomassie staining and silver staining?

Coomassie sensitivity: 50-500 ng protein per band
Silver-staining sensitivity: 1-5 ng protein per band
In general, silver staining is 10-100 times more sensitive than Coomassie staining.

What conditions are expected to remove Coomassie stain from proteins?

The Coomassie dye binds to the protein via hydrophobic interactions, so anything that can disrupt these interactions can potentially serve to remove the dye.

For mild removal conditions, try nonionic detergents such as Triton X-100. If harsher conditions are acceptable, organic solvents like acetonitrile or alcohols should be effective.

Many times, washing with 30% ethanol will remove the stain. Changing the pH or salt concentration is not expected to help.

Are there any suggestions on how to reduce high background when using the Colloidal Blue Staining Kit on Invitrogen Tris-Glycine gels?

It may be possible to reduce background by using the protocol given for NuPAGE Invitrogen Bis-Tris/Small Peptides.

This protocol incorporates an extra fix step to remove excess SDS, which can act as an anti-colloidal agent and lead to higher background.

The low pH of the staining solution will fix the gel, but not as fast as the pre-fix step specified in the NuPAGE protocol.

When staining IEF gels with the Colloidal Blue Staining Kit, is it necessary to use the fixing solution?

Yes, the fix serves two purposes: it fixes the sample in the IEF gel and it helps to remove gel background.

If you do not use the fixing solution, the background on the gels will be high and detection will be less sensitive.

High background is caused by ampholytes remaining in the gel.

Can I stain with Colloidal Blue and then Western blot?

Yes, either Coomassie or Colloidal Blue stain can be used before Western blotting.

However, for optimal transfer efficiency, destain the gel and then equilibrate in a series of Tris base/Glycine/SDS solutions to increase solubility;

When the transfer is complete, the membrane is treated with methanol to remove the stain prior to chromogenic development (not necessary prior to chemilumninescent detection). Review Current Protocols in Molecular Biology Unit 10.8.

Why does a precipitate sometimes form when Colloidal Blue Stainers A and B are mixed together?

The precipitate is caused by a temporary high concentration of one of the components in a localized area.

Gentle mixing of the solution allows the precipitate to go into solution.

What are the blue particles in the Stainer B solution (Colloidal Blue)?

The blue particles are called colloids. They are what enables the kit to work effectively.

Just remember to shake the solution well before using to evenly distribute them throughout the bottle.

Can the Colloidal Blue Staining kit be stored in the refrigerator or freezer?

The Colloidal Blue Staining kit can be stored in the refrigerator, but be sure to let the kit return to room temperature before use.

Also, remember to shake the Stainer B reagent prior to use. Cold storage will not enhance the shelf-life of the stain.

Will the Colloidal Blue Staining Kit stain nucleic acids?

No, due to its chemistry, the Colloidal Blue staining kit only stains proteins and peptides.

Why does the Colloidal Blue Staining Kit appear less intense than standard Coomassie stain?

The Colloidal Blue Staining Kit uses G-250 Coomassie whereas standard Coomassie staining protocols use R-250.

The G-250 is a turquoise color whereas R-250 is closer to navy blue. The difference is in the shade of color, rather than the intensity.

Does the Colloidal Blue Staining kit work with Invitrogen Zymogram gels?

Yes, the Colloidal Blue Staining kit works very well with Invitrogen Zymogram gels.

What reduces precipitation of the Colloidal Blue Staining on gels that are a variation of Ralph Reisfled's "Acid-Urea" gels (6M Urea, 2% Triton X-100, 5% acetic acid), when following the NuPAGE Invitrogen Bis-Tris gel protocol?

These gels are for basic proteins, where they will migrate to the cathode. The interfering substances are urea and Triton X-100.

To improve results, the gels can be rinsed three times (5 min each) with water after the methanol/acetic acid fix step, before adding the staining solution.

How can the Colloidal Blue Staining Kit or SimplyBlue SafeStain be completely removed from the protein for analysis using mass spectrometry?

The G-250 coomassie dye is very soluble in alcohol. To remove all of the stain, incubate the gel in a 10-30% ethanol solution for approximately 15 min or until clear. Rinse and rehydrate the gel in pure water and proceed from there.

In the Colloidal Blue Staining kit manual, it is recommended to incubate the stained gel in GelDry (a 30% alcohol solution) for only 5 min as opposed to the normal 15 min because a 15 min incubation would completely "bleach" the gel.

Can membranes be stained with the Colloidal Blue (G-250) Staining Kit after blotting?

No we do not recommend staining membranes with the Colloidal Blue (G-250) Staining Kit as the background will be too high. Better alternatives include:

- Invitrogen Reversible Membrane Protein Stain (Cat. No. IB7710): Allows for complete, reversible staining of protein on nitrocellulose & PVDF membranes. Sensitivity is higher than Ponceau S (<10 ng of BSA in 10 mins as blue bands) and the staining is reversible in 5 minutes. Western blot detection is unaffected by the staining and erasing process, and in some cases higher sensitivity is achieved.
- Coomassie (non-colloidal) staining: Stain in 0.1% Coomassie Blue R-250 in 50% methanol for 5 minutes and destain with several changes of 50% methanol and 10% acetic acid. Rinse with several changes of water, air dry and store for up to 12 months at −20 degrees C. Sensitivity is approximately at the 50-100 ng level.
- SimplyBlue SafeStain. There is a protocol included in the SimplyBlue SafeStain manual for staining dry PVDF membranes but it is not recommended for wet PVDF membranes or nitrocellulose because of the high background.
- Amido black: same as Coomassie stain but less sensitive. Please see protocol in the manual (https://tools.thermofisher.com/content/sfs/manuals/invitrolonpvdf_man.pdf).
- Ponceau S: same as Coomassie stain but less sensitive. Please see protocol in the manual (https://tools.thermofisher.com/content/sfs/manuals/invitrolonpvdf_man.pdf).
- UV transillumination: After blotting, place the membrane on a filter paper and allow to air-dry at room temperature for about 10 minutes. Rewet in 20% methanol and view the blot in front of white light while it is still wet; the bands will look more translucent than the membrane. If the bands disappear because the membrane is dry, rewet the membrane.

Can I use a pre-stained ladder as a measure of transfer efficiency? Why do I need to use the Invitrogen Reversible Membrane Protein Stain Kit?

The dye that is bound to the proteins in the pre-stained ladder is charged and covalently bound, so the transfer efficiency of pre-stained ladders is almost always better than that of SDS-denatured proteins. Therefore, pre-stained ladders are not a good measure of transfer success. On the other hand, the Invitrogen Reversible Membrane Protein Stain allows for complete, reversible staining of protein on nitrocellulose & PVDF membranes. The staining sensitivity is higher than ponceau S (<10 ng of BSA in 10 mins as blue bands) and the staining is reversible in 5 minutes. Western blot detection is unaffected by the staining and erasing process, and in some cases higher sensitivity is achieved.

Can I stain my gel with Colloidal Blue Stain and then perform western blotting?

Yes, Colloidal Blue Stain can be used before western blotting. However, for optimal transfer efficiency, we recommend destaining the gel and then equilibrating in a series of Tris base/Glycine/SDS solutions to increase solubility; when the transfer is complete, the membrane should be treated with methanol to remove the stain prior to chromogenic development (not necessary prior to chemiluminescent detection).

Why is the background higher on low-percentage acrylamide Tris-Glycine gels after staining with Colloidal Blue?

Background is generally higher in gels less than 10% in acrylamide concentration due to penetration and trapping of colloids within the large pores of the low-percentage acrylamide gels. Excess background may be removed by incubating in 25% methanol solution until a clear background is obtained. Be aware that dye will also be partially removed from the bands and prolonged incubation in >25% methanol will result in a complete destaining of protein bands and background.

By accident, I destained my gel when using the Colloidal Blue Staining kit. Can I restain it?

Yes, you can either completely destain the gel in water and start staining from scratch or if you feel that the staining intensity is a little low and you would like to darken it, you can directly place the gel back in the staining solution.

I am using your Colloidal Blue staining kit and notice a higher amount of colloids than normal. Why is this?

Colloids, or blue chunks, are routine in this type of staining procedure. However, if the amount is abnormally high, it usually indicates that too little or no methanol was added.

I am using the Colloidal Blue Staining kit. I wasn't exact when measuring out the staining solutions; will this affect my results?

Probably not, however if you add greater than 10% less than the total required amount of Stainer B solution, your gel may not stain as intensely as it would have with the proper amount. Adding more of the Stainer B solution will not affect results, nor will +/- 50% of the Stainer A solution.

When Colloidal Blue Stainer A and Stainer B solutions are mixed together, there is a precipitate. Is this normal?

When Colloidal Blue Stainer A and Stainer B solutions are combined, a precipitate may form; this will dissolve within 30 seconds of gentle shaking.

I see blue pieces of material in the Colloidal Blue Stainer B solution. Has it gone bad?

No, the blue “chunks” you see are called colloids. They enable the stain to work effectively. We recommend shaking the solution well before using to evenly distribute these chunks throughout the bottle.

I am using the Colloidal Blue Staining kit and have noticed that the background is higher in low percentage acrylamide gels as compared to high percentage acrylamide gels. Why is this?

- The background is higher in low percentage acrylamide gels due to penetration and trapping of colloids within the large pores of these gels.
- Background may be removed by incubating the gel in 25% methanol solution until a clear background is obtained. Be aware that dye will also be partially removed from the bands.
- Prolonged incubation in >25% methanol results in complete destaining of protein bands and background.

I am getting high background when using the Colloidal Blue Staining kit with Tris-Glycine gels. Can you offer some suggestions?

A reduction in background may be seen if the protocol for NuPAGE Bis-Tris/Small Peptides is used instead. It uses an extra fixing step to remove excess SDS, which can act as an anti-colloidal agent and cause higher background. The low pH of the staining solution will fix the gel, but not as fast as the pre-fixing step performed in the NuPAGE protocol.

Can I use the Colloidal Blue Staining Kit with Invitrogen Zymogram gels?

Yes, the Colloidal Blue Staining kit works very well with Invitrogen Zymogram gels.

What is the difference between Coomassie Blue G250 found in the Colloidal Blue Stain and SimplyBlue SafeStain, and Coomassie R250?

Coomassie G250 is more sensitive than Coomassie R250 whereas Coomassie R250 stains faster than Coomassie G250.

Can I use a gel that is stained with Colloidal Blue Stain for protein sequencing?

Some of the components of the Colloidal Blue Stain are probably too harsh to be used in conjunction with protein sequencing, although the actual Coomassie dye itself does not have an adverse effect.

What wavelengths should I use for densitometry with the Colloidal Blue Coomassie G250 Stain and standard Coomassie R250 stains?

Colloidal Blue Coomassie G250 Stain requires a wavelength of 610 nm whereas standard Coomassie R250 stains require a wavelength of 588 nm.

Can I heat the Colloidal Blue staining solution?

No, heat causes colloids to solubilize; this in turn, causes the free-dye in solution to increase which causes an increase in background and a decrease in staining intensity.

I am using your Colloidal Blue Staining kit. Can I pre-mix the Stainer A and Stainer B solutions and use it at a later date?

No, the staining solution should be prepared fresh, no more than 24 hours before use. If fresh solution is not used, staining background will be high.

When staining with the Colloidal Blue Staining Kit, is it necessary to use the fixing solution for IEF gels?

Yes, the fix serves two purposes: it fixes the sample in the gel and it helps to remove gel background. If you do not use the fixing solution, the background on the gels will be high and detection will be less sensitive. High background can be caused by ampholytes remaining in the gel.

Can I store the Colloidal Blue Staining kit in the refrigerator?

The Colloidal Blue Staining kit can be stored in the refrigerator, but be sure to let the kit return to room temperature before use. Also remember to shake the Stainer B reagent prior to use. Cold storage will not enhance the shelf life of the stain.

Do you have any recommendation for removing Colloidal blue stain from plastic, porcelain and metal surfaces?

A 5% bleach solution will effectively remove the Colloidal Blue stain from plastic, porcelain and metal surfaces.

Do you have any recommendations for drying a gel that has been stained with Colloidal Blue stain?

Please see the following recommendations:

- Do not leave the stained gel in pretreatment gel drying solutions, such as Gel-Dry Solution, for more than 5 minutes.
- Prolonged exposure to pre-treatment gel drying solutions will destain the gel completely.

Can I stain nucleic acids with the Colloidal Blue Staining Kit?

No, due to its chemistry, the Colloidal Blue Staining Kit stains only proteins and peptides. The SilverXpress Staining Kit is a highly sensitive stain for nucleic acids.

Why does staining with the Colloidal Blue Staining Kit appear less intense than with standard Coomassie stain?

The Colloidal Blue Staining Kit uses Coomassie G-250 whereas standard Coomassie staining protocols use Coomassie R-250. Coomassie G-250 is a turquoise color whereas Coomassie R-250 is closer to navy blue. The difference is in the shade of color, rather than the intensity.

Does the Colloidal Blue Staining Kit stain reduced and non-reduced proteins differentially?

Yes, it stains non-reduced proteins more intensely.

Can I stain membranes with the Colloidal Blue (G-250) Staining Kit?

No, we do not recommend staining membranes with the Colloidal Blue (G-250) Staining Kit as the background will be too high. Better alternatives include:

- Coomassie (non-colloidal) staining: Stain in 0.1% Coomassie Blue R-250 in 50% methanol for 5 minutes and destain with several changes of 50% methanol and 10% acetic acid. Rinse with several changes of water, air dry and store for up to 12 months at -20 degrees C. Sensitivity is approximately at the 50-100 ng level.
- SimplyBlue SafeStain. There is a protocol included in the SimplyBlue SafeStain manual for staining PVDF but it is not recommended for nitrocellulose because of the high background.
- Amido Black: same as Coomassie but less sensitive.
- Ponceau S: same as Coomassie but less sensitive.
- UV transillumination: After blotting, place the membrane on a filter paper and allow to dry at room temperature for about 10 minutes. Rewet in 20% methanol and view the blot in front of white light while it is still wet; the bands will look more translucent than the membrane. If the bands disappear because the membrane is dry, rewet the membrane.
- Invitrogen Reversible Membrane Protein Stain (Cat# IB7710): Allows for complete, reversible staining of protein on nitrocellulose & PVDF membranes. Sensitivity is higher than Ponceau S (less than 10 ng of BSA in 10 mins as blue bands) and the staining is reversible in 5 minutes. Western blot detection is unaffected by the staining and erasing process, and in some cases higher sensitivity is achieved.

I would like to send my protein for mass spectrometry (MS) analysis. Can I stain my protein with Colloidal Blue Stain?

Proteins stained using Colloidal Blue Stain are compatible with mass spectrometry (MS) analysis.

What is the sensitivity of detection with the Colloidal Blue Staining kit?

The Colloidal Blue Stain is five times more sensitive than traditional Coomassie Blue staining techniques. Using the Colloidal Blue Staining Kit, less than 10 ng of BSA can be detected on a 4-20% 1.0 mm Invitrogen Tris-Glycine Gel in 1 hour. Non-reduced samples stain slightly more intensely than reduced samples. Bands are visible after 1 hour in staining solution.

How should I store the Colloidal Blue Staining kit and what is its shelf life?

We recommend storing the Colloidal Blue Staining kit at room temperature where it is stable for 1 year.

I stained Tris-Glycine gels with Colloidal Blue stain and noticed that the background was higher in low acrylamide percentage Tris-Glycine gels compared to high acrylamide percentage Tris-Glycine gels. Why is this and how can I resolve it?

Background is generally higher in gels with less than 10% acrylamide percentage due to penetration and trapping of colloids within the large pores of these gels. Excess background may be reduced by incubating the gel in 25% methanol solution until a clear background is obtained. Be aware that the dye will also be partially removed from the bands and that prolonged incubation in >25% methanol will result in complete destaining of protein bands and background.

I used the Colloidal Blue staining kit for staining my Tris-Glycine gels and got very high background. Can you please offer some tips to reduce the background?

It may be possible to reduce background by using the protocol provided for NuPAGE Invitrogen Bis-Tris gels/Small Peptides. This protocol incorporates an extra fix step to remove excess SDS, which can act as an anti-colloidal agent and lead to higher background. The low pH of the staining solution will fix the gel, but not as fast as the pre-fix step specified in the NuPAGE protocol.


For Research Use Only. Not for use in diagnostic procedures.