missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bolt™ Transfer Buffer (20X) DFS Item

Optimized for the transfer of proteins from Bolt Bis-Tris Plus gels to membranes for western blotting

$63.00 - $399.00

Specifications

Content And Storage Store at 4 to 25°C.
Buffer Transfer Buffers
Concentration 20 X
Form Liquid
Gel Compatibility Bolt™ Bis-Tris Plus Gels
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BT0006 DFS Item
View Documents
Invitrogen™
BT0006
125mL
Each for $63.00
Only null left
 
BT00061 DFS Item
View Documents
Invitrogen™
BT00061
1L
Each for $399.00
Only null left
 
Description

Description

  • When combined with 10% methanol, Bolt Transfer Buffer can be used with Bolt Mini Blot Module for wet transfer
  • Can also be used with Bolt Antioxidant to enhance transfer of reduced proteins to membranes

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting

Specifications

Specifications

Store at 4 to 25°C.
20 X
Bolt™ Bis-Tris Plus Gels
Western Blot Transfer Buffer
Transfer Buffers
Liquid
Bolt
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.