Learn More
Invitrogen™ Bolt™ Transfer Buffer (20X)
Description
Requires
A Bolt™ Mini Gel Tank is required for use of all Bolt gels.
- When combined with 10% methanol, Bolt Transfer Buffer can be used with Bolt Mini Blot Module for wet transfer
- Can also be used with Bolt Antioxidant to enhance transfer of reduced proteins to membranes
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
Specifications
| Content And Storage | Store at 4 to 25°C. |
| Buffer | Transfer Buffers |
| Concentration | 20 X |
| Volume (Metric) | 125 ml |
| Form | Liquid |
| Gel Compatibility | Bolt™ Bis-Tris Plus Gels |
| Quantity | 125mL |
| Product Line | Bolt |
| Product Type | Western Blot Transfer Buffer |
Frequently Asked Questions (FAQs)
DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.
Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.
Similar to NuPAGE gels with storage temperatures of 4 to 25 degrees C.
The shelf life for Bolt gels is similar to that for NuPAGE gels: 12 months from the date of shipment.
While they are both Bis-Tris based gels, the chemistries are very different since Bolt gels are optimized for western blotting. Another key difference is the wedge well design of the Bolt gels, which allows larger sample volumes to be loaded.
For Research Use Only. Not for use in diagnostic procedures.