missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Blotting Roller

Catalog No. LC2100
Encompass
Change view
Click to view available options
Quantity:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC2100 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC2100 Supplier Invitrogen™ Supplier No. LC2100
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

One blotting roller

Compatible with E-PAGE™, Novex Mini Gels, and NuPAGE™ Gels

Designed for easy and efficient removal of bubbles when assembling gel/membrane sandwiches for blotting both E-PAGE™ gels and mini-gels such as NuPAGE™ and Novex™ Tris-Glycine gels. Roller is 8.6cm wide and consists of a Delrin roller attached to a stainless steel handle.

Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage One blotting roller.
For Use With (Application) Western Blot
Gel Compatibility E-PAGE™ Gel, Novex™ Mini Gel, NuPAGE™ Gel
Product Line Novex
Type Blotting Roller
Quantity 1 unit
Shipping Condition Room Temperature
What method do you recommend for blotting E-PAGE gels?

The preferred method of transfer for E-PAGE gels is dry blotting using the iBlot/iBlot 2 Dry Blotting System (see Page 27 of the E-PAGE Technical Guide http://tools.thermofisher.com/content/sfs/manuals/epagetechguide_man.pdf). Please note that this works only for E-PAGE 48 gels. Semi-dry blotting using the Invitrogen Semi-Dry Blotter, Cat. No. SD1000 (Page 33), or semi-wet blotting (Page 37) may also be performed.

For Research Use Only. Not for use in diagnostic procedures.