missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BenchMark™ Fluorescent Protein Standard

Consists of fluorescent dye-conjugated proteins, ranging in size from approx. 11-155kDa

Supplier:  Invitrogen™ LC5928

Encompass

Catalog No. LC5928


Only null left

Includes: 125μL (25-50 applications) is provided in loading buffer consisting of Tris-HCl, SDS, Glycerol, and Coomassie Blue G-250

Description

Description

The standard proteins can be resolved into sharp bands in SDS-PAGE and visualized using a UV transilluminator or commonly used laser scanner systems.

 

  • Seven tight, clear bands that resolve well in NuPAGE™ Bis-Tris Gels and Novex Tris-Glycine Gels
  • Visualization directly under the UV light box along with the proteins labeled with the Lumio™ Green Detection Reagent
  • Easy molecular weight estimation for fluorescence-conjugated proteins in SDS-PAGE and western blots
  • A ready-to-use format
  • Detection with Coomassie™ Brilliant Blue R-250
  • Gel Compatibility: NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels

 

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Warranty: 90 days

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

125 μL (25-50 applications) is provided in loading buffer consisting of Tris-HCl, SDS, Glycerol, and Coomassie™ Blue G-250.

Store at -20°C. Avoid repeated freezing and thawing. Stable for 6 months when properly stored.
7
11 to 155 kDa
155, 98, 63, 40, 32, 21, 11 kDa
Dry Ice
Protein Ladder
Western Blotting, SDS-PAGE
488 nm channel
Yes
Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels
125 μL
BenchMark
Alexa Fluor 488
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.