missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ β-Amyloid [42-1] PTD Human Protein

Catalog No. 03247
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03247 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03247 Supplier Invitrogen™ Supplier No. 03247

Inactive control for Aβ [1-42]


  • Format/Formulation: Lyophilized trifluoroacetate salt
  • Molecular Weight: 4515

Shipping Condition: Dry Ice.

Specifications

Gene ID (Entrez) 351
Protein Subtype Neuropeptides, Amyloid-Related Proteins
Content And Storage Store at -5° to -30°C.
For Use With (Application) Cell Analysis; Immunoassays; Ready-To-Use Immunoassay; Build-Your-Own Immunoassay
Quantity 1 mg
Source Human

For Research Use Only. Not for use in diagnostic procedures.