missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 10X Bolt™ Sample Reducing Agent
Optimized for use with Bolt™ Bis-Tris Plus gels, and available in a variety of formats.
$28.12 - $117.00
Specifications
| Content And Storage | Sample Reducing Agent (10X) contains 500 mM dithiothreitol (DTT). Store at 2°C to 8°C. |
|---|---|
| Concentration | 10X, 500 mM |
| Form | Liquid |
| Formulation | 500 mM dithiothreitol (DTT) |
| Gel Compatibility | Bolt™ Bis-Tris Plus Gels |
Includes: 500mM dithiothreitol (DTT)
Description
- Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of small- to medium-sized proteins under denaturing conditions
- Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications
- Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial
- Also use Bolt™ gels to obtain optimal results for day-to-day protein separation needs
- Gels come in four gel types and multiple well formats
- A Mini Gel Tank is required for use of all Bolt™ gels
- Run times may vary depending upon the type of running buffer used
- Store at 2°C to 8°C
Specifications
| Sample Reducing Agent (10X) contains 500 mM dithiothreitol (DTT). Store at 2°C to 8°C. |
|
| Liquid | |
| Bolt™ Bis-Tris Plus Gels | |
| Bolt | |
| Bolt Mini Gel Tank |
| 10X, 500 mM | |
| 500 mM dithiothreitol (DTT) | |
| Reducing Agent | |
| Sample Reducing Agent |
For Research Use Only. Not for use in diagnostic procedures.