Learn More
MilliporeSigma™ Novagen™ Perfect Protein™ Markers, 15-150 kd
Catalog No. 691493
Change view
Click to view available options
Quantity:
100 Lanes
Catalog No. Quantity
691493 100 Lanes
1 options
Catalog No. 691493 Supplier MilliporeSigma™ Supplier No. 691493
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed for routine use in SDS-polyacrylamide gel electrophoresis.
EMD Millipore Novagen™ Perfect Protein Markers, 15-150 kd are set of recombinant proteins with defined sizes at convenient intervals.
  • Optimized for use with Coomassie blue staining
  • Adjusted amounts can also be used with silver staining, fluorescent dyes
  • Includes protein sizes of 15, 25, 35, 50, 75, 100, 150 kd
  • Each vial contains 400μg protein
  • Includes two additional proteins, 10 kd 225 kd, for applications requiring broader size range
  • Each vial contains 500μg protein
Specifications
Quantity 100 Lanes
Storage Requirements −20°C
Product Type DNA Molecular Weight Marker