missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Perfect Protein™ HRP Western Blot Kit

Catalog No. 690783
Change view
Click to view available options
Quantity:
25 Blots
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
690783 25 Blots
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 690783 Supplier MilliporeSigma™ Supplier No. 690783

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detect markers by adding S-protein or His·Tag™ Monoclonal Antibody.

EMD Millipore Novagen™ Perfect Protein HRP Western Blot Kit can be used with colorimetric chemiluminescent AP or HRP substrates.

  • S-protein conjugate does not interfere with antibodies or streptavidin detection
  • Can be used with colorimetric chemiluminescent AP or HRP substrates
  • Recombinant, unmodified markers give sharp, accurately sized bands
  • Markers are supplied at the working dilution in gel loading buffer
  • Concentration optimized for Western detection
  • Protein sizes are 15, 25, 35, 50, 75, 100, 150 kd

Specifications

Description HRP Western Blot Kit
Kit Contents HRP Conjugate, Perfect Protein Western Markers
Quantity 25 Blots
Storage Requirements −20°C
Product Type Protein Transfer/Western Blot Kit