missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Perfect DNA™ 50 bp Ladder

Catalog No. 705383
Change view
Click to view available options
Quantity:
100 Lanes/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
705383 100 Lanes/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 705383 Supplier MilliporeSigma™ Supplier No. MIL705383

Includes

Extra vial of 6X Loading Buffer

Contains sets of DNA for agarose gel analysis.

EMD Millipore Novagen™ Perfect DNA™ 50 bp Ladder has convenient, easy-to-remember sizes.

  • Uniform band intensities
  • Covers wide range of DNA sizes
  • Supplied in convenient ready-to-load format containing dye
  • Suitable for routine use
  • Available in three size ranges: 50 to 3000 bp, 100 to 1000 bp, 500 to 10,000 bp
  • Packaging: Glass Bottle

Specifications

Form Liquid
Quantity 100 Lanes/Pk.
Storage Requirements 2° to 8°C
Product Type DNA Molecular Weight Marker