missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Perfect DNA™ 1 kbp Ladder

Catalog No. 705373
Change view
Click to view available options
Quantity:
100 Lanes/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
705373 100 Lanes/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 705373 Supplier MilliporeSigma™ Supplier No. 705373

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Extra vial of 6X Loading Buffer

Contains sets of DNA for agarose gel analysis.

EMD Millipore Novagen™ Perfect DNA™ 1 kbp Ladder has convenient, easy-to-remember sizes.

  • Uniform band intensities
  • Covers wide range of DNA sizes
  • Supplied in convenient ready-to-load format containing dye
  • Suitable for routine use
  • Available in three size ranges: 50 to 3000 bp, 100 to 1000 bp, 500 to 10,000 bp
  • Packaging: Glass Bottle

Specifications

Form Liquid
Quantity 100 Lanes/Pk.
Storage Requirements 2° to 8°C
Product Type DNA Molecular Weight Marker