missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Novagen™ Overnight Express™ Instant TB Medium
Used as granulated autoinduction culture medium for high-level protein production in pET and other IPTG-inducible bacterial expression systems. Novagen™ Overnight Express™ Instant TB Medium is a granulated medium ensures rapid, uniform dissolution in water, prevents clumping of the medium, and avoids inhalation of airborne powder.
$85.70 - $851.00
Specifications
| Content And Storage | 15°C to 30°C |
|---|---|
| Product Type | Microbiology Media |
| For Use With (Application) | Protein production |
| Packaging Type | Glass Bottle |
Description
- Instant TB Medium is combination of Overnight Express Autoinduction System 1 components (Cat. No. 71300) with rich Terrific Broth (TB) medium and is supplied in two formats
- EasyPak is aluminum foil pouch containing 60g granulated medium, sufficient for 1L culture
- Add the EasyPak contents to 1L sterile water, supplement with 10mL glycerol, and microwave for 2 min to dissolve
- Available in 1kg bottles
Specifications
| 15°C to 30°C | |
| Microbiology Media | |
| Protein production | |
| Glass Bottle |