missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Ni-NTA Buffer Kit

Catalog No. 708993
Change view
Click to view available options
For Use With (Application):
Purification of HisTag fusion proteins
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
708993 Purification of HisTag fusion proteins
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 708993 Supplier MilliporeSigma™ Supplier No. 708993

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for purification of HisTag fusion proteins on Ni-NTA HisBind Resin.

EMD Millipore Novagen™ Ni-NTA Buffer Kit provides convenient set of buffers.

  • Used in HisBind Buffer Kit
  • Prepared 4X concentrates are included for binding, washing elution

Specifications

For Use With (Application) Purification of HisTag fusion proteins