missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ LB Broth, Miller

Used as granulated medium for cultivation of E. coli on scales ranging from small cultures to fermentation. MilliporeSigma™; Novagen™ LB Broth, Miller granules ensure rapid and uniform dissolution in water.

$120.00 - $1470.00

Specifications

Content And Storage 15°C to 30°C
Product Type Microbiology Media
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
717535
View Documents
MilliporeSigma™
717535
500 g
Each for $120.00
Only null left
 
717536
View Documents
MilliporeSigma™
717536
5 kg
Each for $1,470.00
Only null left
 
Description

Description

  • Prevents clumping of medium and inhalation of airborne powder
  • Supplied in several formats
  • EasyPak is aluminum foil pouch containing 25g granulated medium sufficient for 1L culture
  • Add EasyPak contents to 1L water, microwave for 2 minutes to dissolve agar and sterilize
  • Supplied in 500g and 5kg bottles
  • Composition of LB Agar MILLER per liter is 5g yeast extract, 10g peptone from casein, 10g sodium chloride
Specifications

Specifications

15°C to 30°C
Microbiology Media
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only