Learn More
MilliporeSigma™ Novagen™ BLR(DE3) Competent Cells
For production of protein from target genes cloned in pET vectors by induction with IPTG
Supplier:  MilliporeSigma™ 690533
 View more versions of this product
Catalog No. 690533
Only null left
Description
Description
  • recA- derivative of BL21
  • Improves plasmid monomer yields
  • Helps stabilize target plasmids containing repetitive sequences or whose products may cause loss of DE3 prophage (1, 2)
Specifications
Chemical Identifiers
≤ -70°C
Competent Cells
Specifications
≤ -70°C
Competent Cells
0.4mL
Product Suggestions
Product Suggestions