Learn More
MilliporeSigma™ Novagen™, Biotinylated Thrombin
Catalog No. 696723
Change view
Click to view available options
Quantity:
50 U
Catalog No. Quantity
696723 50 U
1 options
Catalog No. 696723 Supplier MilliporeSigma™ Supplier No. 696723
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Removes enzyme from cleavage reactions using immobilized streptavidin.
  • Covalently attached biotin
  • CAS: Ethylenediaminetetraacetic Acid: 60-00-4, Tris Base: 77-86-1
  • Packaging: Glass Bottle
Specifications
Quantity 50 U