missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ B834pLysS Competent Cells

Catalog No. 690424
Change view
Click to view available options
Quantity:
0.4mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
690424 1mL
690423 0.4mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 690424 Supplier MilliporeSigma™ Supplier No. 690424

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For production of protein from target genes cloned in pET vectors by induction with IPTG

  • B834 is parental strain for BL21 (1)
  • Protease-deficient hosts are methionine auxotrophs
  • Allows high specific-activity labeling of target proteins with methionine and selenomethionine for crystallography
  • pLysS strains express T7 lysozyme
  • Suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability

Specifications

Content And Storage ≤ -70°C
Product Type Competent Cells
Quantity 1mL

For Research Use Only