missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ 6X DNA Gel Loading Buffer

Catalog No. 690463
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
690463 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 690463 Supplier MilliporeSigma™ Supplier No. 690463

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Intended for gel electrophoresis of DNA fragments or PCR products.

EMD Millipore Novagen™ 6X DNA Gel Loading Buffer contains Bromophenol Blue, Xylene Cyanol FF, Ficoll.

  • Gel electrophoresis of DNA fragments, PCR products

Specifications

Description 6X DNA Gel Loading Buffer
Color Blue
Concentration 6X
Form Liquid
Quantity 5 mL
Storage Requirements 2° to 8°C
Product Type DNA Sample Loading Buffer