missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nortropinone Hydrochloride 98.0+%, TCI America™
$128.62 - $639.37
Chemical Identifiers
| CAS | 25602-68-0 |
|---|---|
| Molecular Formula | C7H12ClNO |
| Molecular Weight (g/mol) | 161.629 |
| MDL Number | MFCD03613582 |
| InChI Key | MZQWQFWRSDNBPV-UHFFFAOYSA-N |
| Synonym | nortropinone hydrochloride, 8-azabicyclo 3.2.1 octan-3-one hydrochloride, nortropinone hcl, 8-azabicyclo 3.2.1 octan-3-one hcl, 8-azabicyclo 3.2.1 octan-3-one, hydrochloride, 3-nortropanone hydrochloride, 8-aza-3-oxobicyclo 3.2.1 octane hydrochloride, nortropinonehydrochloride, c7h11no.clh, 8-azabicyclo 3,2,1 octan-3-one hydrochloride |
| PubChem CID | 13091218 |
| IUPAC Name | 8-azabicyclo[3.2.1]octan-3-one;hydrochloride |
| SMILES | C1CC2CC(=O)CC1N2.Cl |
Chemical Identifiers
| 25602-68-0 | |
| 161.629 | |
| MZQWQFWRSDNBPV-UHFFFAOYSA-N | |
| 13091218 | |
| C1CC2CC(=O)CC1N2.Cl |
| C7H12ClNO | |
| MFCD03613582 | |
| nortropinone hydrochloride, 8-azabicyclo 3.2.1 octan-3-one hydrochloride, nortropinone hcl, 8-azabicyclo 3.2.1 octan-3-one hcl, 8-azabicyclo 3.2.1 octan-3-one, hydrochloride, 3-nortropanone hydrochloride, 8-aza-3-oxobicyclo 3.2.1 octane hydrochloride, nortropinonehydrochloride, c7h11no.clh, 8-azabicyclo 3,2,1 octan-3-one hydrochloride | |
| 8-azabicyclo[3.2.1]octan-3-one;hydrochloride |
Specifications
| 25602-68-0 | |
| C7H12ClNO | |
| 5 g | |
| MZQWQFWRSDNBPV-UHFFFAOYSA-N | |
| 8-azabicyclo[3.2.1]octan-3-one;hydrochloride | |
| 13091218 | |
| ≥98.0% (T) | |
| Nortropinone Hydrochloride |
| 204°C | |
| MFCD03613582 | |
| nortropinone hydrochloride, 8-azabicyclo 3.2.1 octan-3-one hydrochloride, nortropinone hcl, 8-azabicyclo 3.2.1 octan-3-one hcl, 8-azabicyclo 3.2.1 octan-3-one, hydrochloride, 3-nortropanone hydrochloride, 8-aza-3-oxobicyclo 3.2.1 octane hydrochloride, nortropinonehydrochloride, c7h11no.clh, 8-azabicyclo 3,2,1 octan-3-one hydrochloride | |
| C1CC2CC(=O)CC1N2.Cl | |
| 161.629 | |
| 161.63 | |
| Crystalline Powder |
Safety and Handling
TSCA : No