missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ North™ NorthFlex Red™ Foam PVC Coated Nylon Gloves
Description
- Foam PVC coating on the palm for enhanced grip capabilities
- Seamless nylon shell provides a snug fit for comfort and safety
- Ventilated, uncoated glove back keeps hands cool
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 4 |
| Certifications/Compliance | ANSI 4 |
| Coated Area | Palm, Fingers |
| Construction | Seamless Knit |
| Coating Material | PVC |
| Color | Red |
| Hand | Specific |
| Material | Nylon |
| Product Type | Seamless Knit Gloves |
| Size | X-Large |
| Show More |