Learn More
Bullard™ Hardhat and Cap Face Shield Aluminum Brackets
Catalog No. 17360
Change view
Click to view available options
For Use With (Equipment):
C30 Series Cap-style Hard Hat Models, Faceshield
C30, S61, S62, S51, 911C Cap-style, 911H Full Brim Series and Bump Cap Type II Fard Hat, Faceshield
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17360 C30 Series Cap-style Hard Hat Models, Faceshield
19021320 C30, S61, S62, S51, 911C Cap-style, 911H Full Brim Series and Bump Cap Type II Fard Hat, Faceshield
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17360 Supplier Bullard™ Supplier No. 500M
Rugged brackets are ideal for high-heat industrial applications
Aluminum Brackets: Gold Line™ brackets are constructed of rugged, gold anodized aluminum. Highly resistant to corrosion and heat. Ideal for chemical handling and high heat industrial applications. Band- or slot-mounted.
Specifications
For Use With (Equipment) C30 Series Cap-style Hard Hat Models, Faceshield