Learn More
Splash Shield™ North™ Protecto-Shield Face Shields
Catalog No. 17310
Change view
Click to view available options
Disposable Face Shield
Catalog No.
17310 Disposable Face Shield
1 options
Catalog No. 17310 Supplier Splash Shield™ Supplier No. 4505B
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Fits well over glasses and protective spectacles
  • Disposable
  • Constructed from high-quality optically clear, distortion-free 7mil polyester film
  • Headpiece is held in place by stretch strap
Compliance
Conforms to OSHA 29CFR Part 1910.1030 for bloodborne pathogens and CDC recommendation for protective eyewear.
Specifications
Lens Tint Clear
Product Type Splash Shield
For Use With (Application) Fits well over glasses and protective spectacles
Material Polyester
Disposable Single-use