Learn More
Honeywell™ North™ ER2000CBRN Escape Respirator
Catalog No. 191501568
Change view
Click to view available options
Escape respirator; ER2000
Catalog No.
191501568 Escape respirator; ER2000
1 options
Catalog No. 191501568 Supplier Honeywell™ Supplier No. ER2000CBRN
Easy and intuitive to use
Hood style respirator provides respiratory protection from Chemical, Biological, Radiological and Nuclear (CBRN) warfare agents as well as Toxic Industrial Materials (TIM's). Comfortable hood and can be donned and functional within seconds.
  • Highly reflective head strap harness ensures user visibility and a comfortable and secure fit
  • Utilizes proven half-mask design for optimum comfort and communication ability
  • Cartridges contain impregnated ASZM-TEDA carbon as specified for US Military canisters, for protection from chemical war agents
  • HEPA filter is sealed to the cartridge for particulate protection
  • Universal size
Compliance
NIOSH/CBRN approval TC-14G-028. Meets CAP 2, 30 minute rating.
Specifications
Certifications/Compliance NIOSH CBRN TC-14G-028, CAP 2
Product Type Escape Respirator
For Use With (Application) Provides respiratory protection from chemical, biological, radiological and nuclear warfare agents as well as toxic industrial material
Includes Hood with a neck seal, an oral, nasal cup and two air purifying chemical, biological, radiological and nuclear cartridge, anti-fog and scratch resistant lens, highly reflective head strap harness, resealable foil package, hepa filter
Weight (Metric) 0.6 kg
Size Universal
Dimensions (L x W x H) 11.5 x 9.5 x 8 in. (29.21 x 24.13 x 20.32 cm)
Weight (English) 1.33 lb.