Learn More
Honeywell™ North™ Compact Air PAPR Hood
Catalog No. 19134249
Change view
Click to view available options
For Use With (Equipment):
Compact air 200 series powered air purifying respirator and cf1000 series
Catalog No. For Use With (Equipment)
19134249 Compact air 200 series powered air purifying respirator and cf1000 series
1 options
Catalog No. 19134249 Supplier Honeywell™ Supplier No. PA112
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Replacement hood for Primair™ 100 Bibbed Hood Assembly that can be used with the CA200 Series PAPR and the CF1000 Series CF-SAR.
  • Tychem QC hood with full bib
  • Hoods have an assigned protection factor (APF) of 1000
  • NIOSH certified when used with North Compact Air PAPR, CF1000 supplied air assembly, and North hoses and couplers
Specifications
Certifications/Compliance NIOSH
Material Tyvek
Product Type Respirator Hood
For Use With (Application) Painting applications and other environments where full head and shoulder coverage is desired
For Use With (Equipment) Compact air 200 series powered air purifying respirator and cf1000 series
Includes Hood, adjustable headgear, replacement sweatband and 1 set peel-away windows
Model PA112