missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ 5500 Series Half-Mask Air Purifying Respirators

Catalog No. 18987A
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18987A Small
18987B Medium
18987C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18987A Supplier Honeywell™ Supplier No. 550030 S

Incorporates the low-profile design of the North 7700 Series in an economical material. Cartridges and filters not included.

  • Easy-to-clean facepiece conforms gently to wearer's face and is made from a soft elastomer that resists distortion
  • 'One-piece' cradle suspension system is easily removed for cleaning and provides a comfortable, secure fit with no slipping
  • Contoured sealing flange eliminates pressure points
  • Extended side flanges ensure secure seal even when wearer is talking
  • Exhalation valve assembly provides low breathing resistance and permits easy, positive pressure fit check without removing cover
  • Headband straps are adjustable and virtually impossible to lose or misassemble; headband yoke improves peripheral vision by allowing cartridges to be lower and farther back
  • Direct cartridge-to-facepiece seal eliminates risk of improper seal and reduced protection due to lost or worn sealing gaskets
  • Requires limited maintenance and available in three sizes that accept North cartridges and filters

Compliance

NIOSH approved under 42 CFR Part 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters.

Specifications

Certifications/Compliance MSHA
Certifications (NIOSH) NIOSH 42 CFR Part 84
Certifications (OSHA) OSHA 29 CFR Part 1920.134
Connection Type Threaded
Color Gray
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small
For Use With (Equipment) North™ Cartridge, Filter and Accessories
Model 550030S