missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ North™ 5400 Series Full Facepiece Air-Purifying Respirator

Catalog No. 19025040A
Change view
Click to view available options
Size:
Medium/Large
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19025040A Small
19025040B Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19025040A Supplier Honeywell™ Supplier No. 54001 S

Requires

Compatible with North cartridges, filters and accessories.

Full face 5400 series are one of the lightest respirators on the market and can be used as part of a PAPR or supplied air system. Cartridges and filters sold separately.

  • Face seal and nose cup are made from a thermoplastic elastomer (TPE)
  • Soft, pliable, lightweight material resists alcohol, acids and bases
  • Anti-scratch polycarbonate wide panoramic lens provides optimal field of vision in all directions
  • Oral/nasal cup minimizes lens fogging
  • Threaded cartridge connector secures equipment to avoid accidental disconnections
  • 5400 respirator is compatible with all North cartridges, filters, and accessories
  • Can be adapted to PAPR or CF-SAR; compatible with PA700 Series PAPR and CF2000 Series SAR

Compliance

NIOSH approved under 42 CFR 84 when used as designed with appropriate cartridges and filters.

Specifications

ANSI Standard ANSI Z87.1
Certifications (NIOSH) NIOSH 42 CFR Part 84
Certifications (OSHA) OSHA 29 CFR Part 1920.134
Connection Type Threaded
Color Black
Material Thermoplastic Elastomer
Product Type Full Facepiece Respirator
Size Small
For Use With (Equipment) North™ N-Series line of cartridges, filters, and cartridge/filter combinations
Material (Lens) Polycarbonate
Model 54001S
Suspension Type 4-point
Weight (English) 14 lb.
Weight (Metric) 6.4 kg
Show More Show Less